2-Chloro-1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

318606-1G 318606-500MG 318606-5G
77469-058EA 696.28 USD
77469-058 77481-734 77492-128
2-Chloro-1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₅ClF₂O₃S
MW: 254.6383 g/mol
Storage Temperature: Ambient
CAS Number: 2029838-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR