1-(3,5-dimethyl-4-propoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

318600-1G 318600-500MG 318600-5G
77469-054EA 284.28 USD
77469-054 77481-730 77492-124
1-(3,5-dimethyl-4-propoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(3,5-dimethyl-4-propoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₆H₂₆O₂
MW: 250.3764 g/mol
Storage Temperature: Ambient
CAS Number: 3002532-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR