(2-Bromo-6-fluoro-3-propoxyphenyl)(5-methylthiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

318598-1G 318598-500MG 318598-5G
77469-050EA 415.09 USD
77469-050 77481-726 77492-120
(2-Bromo-6-fluoro-3-propoxyphenyl)(5-methylthiophen-2-yl)methanol ≥95%
(2-Bromo-6-fluoro-3-propoxyphenyl)(5-methylthiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆BrFO₂S
Storage Temperature: Ambient
CAS Number: 3002532-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR