1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

318594-1G 318594-500MG 318594-5G
77469-046EA 415.09 USD
77469-046 77481-722 77492-116
1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylbutan-1-ol ≥95%
1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₀BrFO₂
MW: 319.2098 g/mol
Storage Temperature: Ambient
CAS Number: 3002475-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR