(2,6-Difluoro-4-methoxyphenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

318582-1G 318582-500MG 318582-5G
77469-042EA 415.09 USD
77469-042 77481-718 77492-112
(2,6-Difluoro-4-methoxyphenyl)(4-ethylphenyl)methanol ≥95%
(2,6-Difluoro-4-methoxyphenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₆F₂O₂
MW: 278.2938 g/mol
Storage Temperature: Ambient
CAS Number: 1292106-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR