1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

318577-1G 318577-500MG 318577-5G
77469-038EA 415.09 USD
77469-038 77481-714 77492-108
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₇BrF₄O
MW: 299.0595 g/mol
Storage Temperature: Ambient
CAS Number: 3002535-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR