(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

318575-1G 318575-500MG 318575-5G
77469-034EA 415.09 USD
77469-034 77481-710 77492-104
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₃BrF₄O
Storage Temperature: Ambient
CAS Number: 3002484-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR