α-Cyclopropyl-2-methyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

317936-1G 317936-500MG 317936-5G
77469-026EA 415.09 USD
77469-026 77481-702 77492-096
α-Cyclopropyl-2-methyl-1-naphthalenemethanol ≥95%
α-Cyclopropyl-2-methyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₅H₁₆O
Storage Temperature: Ambient
CAS Number: 1537857-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR