Cyclobutyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

317031-1G 317031-500MG 317031-5G
77469-022EA 546.93 USD
77469-022 77481-698 77492-092
Cyclobutyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone ≥95%
Cyclobutyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone

Precaución: For research use only

Formula: C₁₂H₁₀F₄O
MW: 246.2 g/mol
Storage Temperature: Ambient
CAS Number: 1774825-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR