1-(2-Bromo-3-chloro-6-fluorophenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

316744-1G 316744-500MG 316744-5G
77469-018EA 415.09 USD
77469-018 77481-694 77492-088
1-(2-Bromo-3-chloro-6-fluorophenyl)-2-methylbutan-1-ol ≥95%
1-(2-Bromo-3-chloro-6-fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.5757 g/mol
Storage Temperature: Ambient
CAS Number: 3002483-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR