(2-Bromo-3-chloro-6-fluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

316740-1G 316740-500MG 316740-5G
77469-010EA 415.09 USD
77469-010 77481-686 77492-080
(2-Bromo-3-chloro-6-fluorophenyl)triethylsilane ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₇BrClFSi
MW: 323.7042 g/mol
Storage Temperature: Ambient
CAS Number: 3002474-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR