(2-Bromo-3-chloro-6-fluorophenyl)(cyclopropyl)methanol ≥97%

Supplier: AOBChem USA

Warning

316738-1G 316738-500MG 316738-5G
77469-006EA 428.48 USD
77469-006 77481-682 77492-076
(2-Bromo-3-chloro-6-fluorophenyl)(cyclopropyl)methanol ≥97%
(2-Bromo-3-chloro-6-fluorophenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₀H₉BrClFO
Storage Temperature: Ambient
CAS Number: 3002469-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR