1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

316704-1G 316704-500MG 316704-5G
77469-002EA 415.09 USD
77469-002 77481-676 77492-070
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)propan-1-ol ≥95%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₆H₁₆BrFO₂
MW: 339.1994 g/mol
Storage Temperature: Ambient
CAS Number: 3002482-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR