1-(Benzyloxy)-2,3-dibromo-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

316699-1G 316699-500MG 316699-5G
77468-994EA 415.09 USD
77468-994 77481-670 77492-064
1-(Benzyloxy)-2,3-dibromo-4-fluorobenzene ≥95%
1-(Benzyloxy)-2,3-dibromo-4-fluorobenzene

Precaución: For research use only

Formula: C₁₃H₉Br₂FO
MW: 360.0163 g/mol
Storage Temperature: Ambient
CAS Number: 3002474-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR