1-Bromo-4-ethoxy-2-fluoro-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

31646-1G 31646-500MG 31646-5G
77468-986EA 258.53 USD
77468-986 77481-662 77492-056
1-Bromo-4-ethoxy-2-fluoro-5-methoxybenzene ≥97%
1-Bromo-4-ethoxy-2-fluoro-5-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28401602
CAS Number: 1095544-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR