4-Butyl-4'-(ethylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

315285-1G 315285-500MG 315285-5G
77468-982EA 415.09 USD
77468-982 77481-658 77492-050
4-Butyl-4'-(ethylthio)-1,1'-biphenyl ≥95%
4-Butyl-4'-(ethylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₈H₂₂S
MW: 270.43 g/mol
Storage Temperature: Ambient
CAS Number: 104560-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR