B-(4'-Butyl[1,1'-biphenyl]-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

315281-1G 315281-500MG 315281-5G
77468-978EA 835.33 USD
77468-978 77481-654 77492-046
B-(4'-Butyl[1,1'-biphenyl]-4-yl)boronic acid ≥95%
B-(4'-Butyl[1,1'-biphenyl]-4-yl)boronic acid

Precaución: For research use only

Formula: C₁₆H₁₉BO₂
MW: 254.13 g/mol
Storage Temperature: Ambient
CAS Number: 145413-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR