4-(4-Methoxyphenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Methoxyphenyl)pyridine

314752-1G 314752-500MG 314752-5G
77468-974EA 77.25 USD
77468-974 77481-650 77492-042
4-(4-Methoxyphenyl)pyridine ≥95%
4-(4-Methoxyphenyl)pyridine

Precaución: For research use only

Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092331
CAS Number: 5938-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR