3-(Benzyloxy)-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

31455-1G 31455-500MG 31455-5G
77468-970EA 284.28 USD
77468-970 77481-646 77492-038
3-(Benzyloxy)-5-(trifluoromethyl)benzoic acid ≥95%
3-(Benzyloxy)-5-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₁₅H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD11848448
CAS Number: 53985-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR