1,3-Dimethoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

31450-1G 31450-5G
77468-966EA 71.07 USD
77468-966 77492-034
1,3-Dimethoxy-5-(trifluoromethyl)benzene ≥97%
1,3-Dimethoxy-5-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18395741
CAS Number: 158152-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR