(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314286-1G 314286-500MG 314286-5G
77468-958EA 415.09 USD
77468-958 77481-640 77492-026
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(propyl)sulfane ≥95%
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₁BrClFS
Storage Temperature: Ambient
CAS Number: 3002427-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR