(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

314278-1G 314278-500MG 314278-5G
77468-946EA 415.09 USD
77468-946 77481-628 77492-014
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclopropyl)methanol ≥95%
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₁BrClFO
MW: 293.5598 g/mol
Storage Temperature: Ambient
CAS Number: 3002541-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR