(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314276-1G 314276-500MG 314276-5G
77468-942EA 415.09 USD
77468-942 77481-624 77492-010
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(ethyl)sulfane ≥95%
(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₉BrClFS
Storage Temperature: Ambient
CAS Number: 3002530-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR