1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

314273-1G 314273-500MG 314273-5G
77468-938EA 415.09 USD
77468-938 77481-620 77492-006
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-fluoroethanone ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₉H₆BrClF₂O
MW: 283.4971 g/mol
Storage Temperature: Ambient
CAS Number: 3002527-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR