1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

314271-1G 314271-500MG 314271-5G
77468-934EA 415.09 USD
77468-934 77481-616 77492-002
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Bromo-3-chloro-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₄BrClF₄O
MW: 319.4780 g/mol
Storage Temperature: Ambient
CAS Number: 3002429-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR