(6-Bromo-2-fluoro-3-propoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314265-1G 314265-500MG 314265-5G
77468-930EA 415.09 USD
77468-930 77481-612 77491-998
(6-Bromo-2-fluoro-3-propoxyphenyl)(propyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₆BrFOS
Storage Temperature: Ambient
CAS Number: 3002473-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR