1-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

314263-1G 314263-500MG 314263-5G
77468-926EA 415.09 USD
77468-926 77481-608 77491-994
1-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.1566 g/mol
Storage Temperature: Ambient
CAS Number: 3002512-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR