1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

314261-1G 314261-500MG 314261-5G
77468-922EA 415.09 USD
77468-922 77481-604 77491-990
1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₂H₉BrF₆O₂
MW: 379.0930 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR