2-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

314257-1G 314257-500MG 314257-5G
77468-918EA 415.09 USD
77468-918 77481-600 77491-986
2-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-2-ol ≥95%
2-(6-Bromo-2-fluoro-3-propoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.1566 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR