(2-Bromo-3-ethoxy-6-fluorophenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314242-1G 314242-500MG 314242-5G
77468-914EA 415.09 USD
77468-914 77481-596 77491-982
(2-Bromo-3-ethoxy-6-fluorophenyl)(isopropyl)sulfane ≥95%
(2-Bromo-3-ethoxy-6-fluorophenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₄BrFOS
Storage Temperature: Ambient
CAS Number: 3002487-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR