(6-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

314233-1G 314233-500MG 314233-5G
77468-910EA 415.09 USD
77468-910 77481-592 77491-978
(6-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₂BrFOS
MW: 279.1690 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR