1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

314228-1G 314228-500MG 314228-5G
77468-902EA 415.09 USD
77468-902 77481-584 77491-970
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.1566 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR