1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

314226-1G 314226-500MG 314226-5G
77468-898EA 415.09 USD
77468-898 77481-580 77491-966
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O₂
MW: 317.4870 g/mol
Storage Temperature: Ambient
CAS Number: 3002512-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR