2-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

314224-1G 314224-500MG 314224-5G
77468-894EA 573.71 USD
77468-894 77481-576 77491-962
2-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol ≥95%
2-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO₂
MW: 263.1034 g/mol
Storage Temperature: Ambient
CAS Number: 1779748-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR