1-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

314220-1G 314220-500MG 314220-5G
77468-890EA 415.09 USD
77468-890 77481-572 77491-958
1-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-methoxyphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO₂
MW: 263.1034 g/mol
Storage Temperature: Ambient
CAS Number: 3002429-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR