2-Methylquinoline-6-boronic acid ≥95%

Supplier: AOBChem USA

Warning

31321-1G 31321-500MG 31321-5G
77468-886EA 268.83 USD
77468-886 77481-568 77491-954
2-Methylquinoline-6-boronic acid ≥95%
2-Methylquinoline-6-boronic acid

Precaución: For research use only

Formula: C₁₀H₁₀BNO₂
MW: 187.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062388
CAS Number: 1092790-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR