(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

313169-1G 313169-500MG 313169-5G
77468-882EA 415.09 USD
77468-882 77481-564 77491-950
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclohexyl)methanol ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₅BrF₄O
Storage Temperature: Ambient
CAS Number: 3002429-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR