1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

313167-1G 313167-500MG 313167-5G
77468-878EA 415.09 USD
77468-878 77481-560 77491-946
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₉BrF₄O
MW: 301.0754 g/mol
Storage Temperature: Ambient
CAS Number: 3002477-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR