(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

313161-1G 313161-500MG 313161-5G
77468-870EA 415.09 USD
77468-870 77481-552 77491-938
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₉BrF₄O
MW: 313.0861 g/mol
Storage Temperature: Ambient
CAS Number: 3002436-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR