(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

313159-1G 313159-500MG 313159-5G
77468-866EA 415.09 USD
77468-866 77481-548 77491-934
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₇BrF₄S
Storage Temperature: Ambient
CAS Number: 3002524-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR