Isopropyl(4-methylnaphthalen-1-yl)sulfane ≥95%

Supplier: AOBChem USA

Warning

313132-1G 313132-500MG 313132-5G
77468-862EA 415.09 USD
77468-862 77481-544 77491-930
Isopropyl(4-methylnaphthalen-1-yl)sulfane ≥95%
Isopropyl(4-methylnaphthalen-1-yl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₆S
MW: 216.3418 g/mol
Storage Temperature: Ambient
CAS Number: 3002475-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR