2-Chloro-2,2-difluoro-1-(4-methylnaphthalen-1-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

313127-1G 313127-500MG 313127-5G
77468-858EA 543.84 USD
77468-858 77481-540 77491-926
2-Chloro-2,2-difluoro-1-(4-methylnaphthalen-1-yl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(4-methylnaphthalen-1-yl)ethanone

Precaución: For research use only

Formula: C₁₃H₉ClF₂O
MW: 254.6597 g/mol
Storage Temperature: Ambient
CAS Number: 2016853-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR