[1,1'-biphenyl]-2-yl(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

313120-1G 313120-500MG 313120-5G
77468-854EA 415.09 USD
77468-854 77481-536 77491-922
[1,1'-biphenyl]-2-yl(ethyl)sulfane ≥95%
[1,1'-biphenyl]-2-yl(ethyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₄S
Storage Temperature: Ambient
CAS Number: 1416789-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR