1-(5-Methoxybenzo[d][1,3]dioxol-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

312961-1G 312961-500MG 312961-5G
77468-850EA 600.49 USD
77468-850 77481-532 77491-918
1-(5-Methoxybenzo[d][1,3]dioxol-4-yl)ethanone ≥95%
1-(5-Methoxybenzo[d][1,3]dioxol-4-yl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Ambient
CAS Number: 1780638-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR