6-Chloro-2-fluoropyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31255-1G 31255-500MG 31255-5G
77468-846EA 426.42 USD
77468-846 77481-528 77491-914
6-Chloro-2-fluoropyridine-3-boronic acid pinacol ester ≥97%
6-Chloro-2-fluoropyridine-3-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₁H₁₄BClFNO₂
MW: 257.497 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182150
CAS Number: 1628524-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR