1-Bromo-5-chloro-2-fluoro-4-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

31202-1G 31202-500MG 31202-5G
77468-842EA 282.22 USD
77468-842 77481-524 77491-908
1-Bromo-5-chloro-2-fluoro-4-isopropoxybenzene ≥95%
1-Bromo-5-chloro-2-fluoro-4-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088276
CAS Number: 1879026-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR