5-Chloro-2-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31191-1G 31191-500MG 31191-5G
77468-838EA 224.54 USD
77468-838 77481-520 77491-904
5-Chloro-2-fluoro-4-methoxybenzaldehyde ≥97%
5-Chloro-2-fluoro-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06246209
CAS Number: 221622-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR