1-Bromo-5-chloro-2-fluoro-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

31172-1G 31172-500MG 31172-5G
77468-834EA 157.59 USD
77468-834 77481-516 77491-900
1-Bromo-5-chloro-2-fluoro-4-(methoxymethoxy)benzene ≥95%
1-Bromo-5-chloro-2-fluoro-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrClFO₂
MW: 269.495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619428
CAS Number: 1872221-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR