Ethyl 4-bromo-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

31163-1G 31163-500MG 31163-5G
77468-830EA 282.22 USD
77468-830 77481-512 77491-898
Ethyl 4-bromo-2,5-dimethylbenzoate ≥97%
Ethyl 4-bromo-2,5-dimethylbenzoate

Precaución: For research use only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18905463
CAS Number: 189208-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR