3,5-Dibromo-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

31048-1G 31048-500MG 31048-5G
77468-822EA 140.08 USD
77468-822 77481-504 77491-888
3,5-Dibromo-4-(methylthio)pyridine ≥97%
3,5-Dibromo-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅Br₂NS
MW: 282.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30174232
CAS Number: 1357094-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR